Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250035.2 | XP_008248257.1 | 157 | 99 | ||
| XM_008250041.2 | XP_008248263.1 | 157 | 99 | |||
| XM_008250043.2 | XP_008248265.1 | 154 | 99 | |||
| XM_017338448.1 | XP_017193937.1 | 154 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008250035.2, XM_008250041.2, XM_008250043.2, XM_017338448.1 transcripts.
Set Membership: |
|

ZNF248
Genomic Map