Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337978.1 | XP_017193467.1 | 119 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017337978.1 transcripts.
Set Membership: |
|

CD209
Genomic Map