Assay Details
| Gene Symbol: CD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340148.1 | XP_017195637.1 | 351 | 55 | ||
| XM_017340149.1 | XP_017195638.1 | 422 | 55 | |||
| XM_017340150.1 | XP_017195639.1 | 364 | 55 | |||
More Information
Set Membership: |
|

CD37
Genomic Map