Assay Details
| Gene Symbol: NAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710936.3 | XP_002710982.1 | 2 - 3 | 276 | 65 | |
| XM_017341770.1 | XP_017197259.1 | 2 - 3 | 280 | 65 | ||
| XM_017341772.1 | XP_017197261.1 | 2 - 3 | 283 | 65 | ||
| XM_017341773.1 | XP_017197262.1 | 1 - 2 | 78 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017341773.1 transcripts.
Set Membership: |
|

NAPB
Genomic Map