Assay Details
| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340139.1 | XP_017195628.1 | 1 - 2 | 104 | 103 | |
| XM_017340141.1 | XP_017195630.1 | 1 - 2 | 104 | 103 | ||
More Information
Set Membership: |
|

IL37
Genomic Map