Assay Details
Gene Symbol: TSPAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712109.3 | XP_002712155.1 | 6 - 7 | 619 | 78 | |
XM_008258287.2 | XP_008256509.1 | 7 - 8 | 821 | 78 | ||
XM_017342721.1 | XP_017198210.1 | 6 - 7 | 841 | 78 | ||
XM_017342722.1 | XP_017198211.1 | 6 - 7 | 747 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
