Assay Details
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268110.2 | XP_008266332.1 | 3 - 4 | 630 | 65 | |
XM_008268111.2 | XP_008266333.1 | 3 - 4 | 630 | 65 | ||
XM_008268112.2 | XP_008266334.1 | 3 - 4 | 620 | 65 | ||
XM_008268113.2 | XP_008266335.1 | 3 - 4 | 630 | 65 | ||
XM_008268114.2 | XP_008266336.1 | 3 - 4 | 630 | 65 | ||
XM_008268115.2 | XP_008266337.1 | 3 - 4 | 630 | 65 | ||
XM_008268116.2 | XP_008266338.1 | 3 - 4 | 630 | 65 | ||
XM_008268117.2 | XP_008266339.1 | 3 - 4 | 630 | 65 | ||
XM_008268118.2 | XP_008266340.1 | 3 - 4 | 630 | 65 | ||
XM_017347690.1 | XP_017203179.1 | 2 - 3 | 546 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
