Assay Details
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008260752.2 | XP_008258974.1 | 5 - 6 | 741 | 61 | |
XM_008260753.2 | XP_008258975.1 | 5 - 6 | 750 | 61 | ||
XM_008260754.2 | XP_008258976.1 | 5 - 6 | 744 | 61 | ||
XM_008260756.2 | XP_008258978.1 | 5 - 6 | 738 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
