Assay Details
| Gene Symbol: LOC100356626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259878.2 | XP_008258100.1 | 7 - 8 | 1009 | 70 | |
| XM_008259879.2 | XP_008258101.1 | 7 - 8 | 1009 | 70 | ||
| XM_017343450.1 | XP_017198939.1 | 7 - 8 | 1010 | 70 | ||
More Information
Set Membership: |
|

LOC100356626
Genomic Map