Assay Details
| Gene Symbol: PHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712768.3 | XP_002712814.2 | 2 - 3 | 496 | 62 | |
| XM_008259796.2 | XP_008258018.1 | 3 - 4 | 505 | 62 | ||
| XM_008259797.2 | XP_008258019.1 | 3 - 4 | 505 | 62 | ||
| XM_008259798.2 | XP_008258020.1 | 3 - 4 | 505 | 62 | ||
| XM_017343426.1 | XP_017198915.1 | 3 - 4 | 505 | 62 | ||
More Information
Set Membership: |
|

PHC1
Genomic Map