Assay Details
| Gene Symbol: RAET1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249898.2 | XP_008248120.1 | 1041 | 90 | ||
| XM_017338347.1 | XP_017193836.1 | 1074 | 90 | |||
| XM_017338348.1 | XP_017193837.1 | 1039 | 90 | |||
| XM_017338349.1 | XP_017193838.1 | 1173 | 90 | |||
| XM_017338350.1 | XP_017193839.1 | 898 | 90 | |||
More Information
Set Membership: |
|

RAET1E
Genomic Map