Assay Details
Gene Symbol: C13H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264672.2 | XP_008262894.1 | 2 - 3 | 514 | 81 | |
XM_008264673.2 | XP_008262895.1 | 2 - 3 | 593 | 81 | ||
XM_008264674.2 | XP_008262896.1 | 2 - 3 | 518 | 81 | ||
XM_008264675.2 | XP_008262897.1 | 3 - 4 | 501 | 81 | ||
XM_008264676.2 | XP_008262898.1 | 3 - 4 | 422 | 81 | ||
XM_008264677.2 | XP_008262899.1 | 3 - 4 | 472 | 81 | ||
XM_008264678.2 | XP_008262900.1 | 3 - 4 | 393 | 81 | ||
XM_017345984.1 | XP_017201473.1 | 3 - 4 | 426 | 81 | ||
XM_017345985.1 | XP_017201474.1 | 2 - 3 | 354 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
