Assay Details
| Gene Symbol: C13H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264672.2 | XP_008262894.1 | 4 - 5 | 681 | 90 | |
| XM_008264673.2 | XP_008262895.1 | 4 - 5 | 760 | 90 | ||
| XM_008264674.2 | XP_008262896.1 | 4 - 5 | 685 | 90 | ||
| XM_008264675.2 | XP_008262897.1 | 5 - 6 | 668 | 90 | ||
| XM_008264676.2 | XP_008262898.1 | 5 - 6 | 589 | 90 | ||
| XM_008264677.2 | XP_008262899.1 | 5 - 6 | 639 | 90 | ||
| XM_008264678.2 | XP_008262900.1 | 5 - 6 | 560 | 90 | ||
| XM_017345984.1 | XP_017201473.1 | 5 - 6 | 593 | 90 | ||
| XM_017345985.1 | XP_017201474.1 | 4 - 5 | 521 | 90 | ||
More Information
Set Membership: |
|

C13H1orf162
Genomic Map