Assay Details
| Gene Symbol: BICD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708227.3 | XP_002708273.3 | 6 - 7 | 2347 | 63 | |
| XM_017342054.1 | XP_017197543.1 | 6 - 7 | 2347 | 63 | ||
More Information
Set Membership: |
|

BICD2
Genomic Map