Assay Details
| Gene Symbol: TXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713546.3 | XP_002713592.1 | 6 - 7 | 870 | 78 | |
| XM_008261338.2 | XP_008259560.1 | 6 - 7 | 872 | 78 | ||
| XM_017344200.1 | XP_017199689.1 | 6 - 7 | 872 | 78 | ||
More Information
Set Membership: |
|

TXNL1
Genomic Map