Assay Details
| Gene Symbol: BCL2L11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340061.1 | XP_017195550.1 | 4 - 5 | 620 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL2L11| Gene Symbol: BCL2L11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340061.1 | XP_017195550.1 | 4 - 5 | 620 | 68 | |
Set Membership: |
|