Assay Details
| Gene Symbol: CBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271355.2 | XP_008269577.1 | 4 - 5 | 671 | 64 | |
| XM_008271356.2 | XP_008269578.1 | 4 - 5 | 471 | 64 | ||
| XM_008271358.2 | XP_008269580.1 | 4 - 5 | 545 | 64 | ||
| XM_017349170.1 | XP_017204659.1 | 5 - 6 | 843 | 64 | ||
More Information
Set Membership: |
|

CBX1
Genomic Map