Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711901.3 | XP_002711947.1 | 6 - 7 | 1437 | 64 | |
| XM_008257972.2 | XP_008256194.1 | 7 - 8 | 1633 | 64 | ||
| XM_008257973.2 | XP_008256195.1 | 7 - 8 | 1492 | 64 | ||
| XM_008257974.2 | XP_008256196.1 | 6 - 7 | 1491 | 64 | ||
| XM_008257976.2 | XP_008256198.1 | 6 - 7 | 1472 | 64 | ||
More Information
Set Membership: |
|

DTX2
Genomic Map