Assay Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259742.2 | XP_008257964.1 | 10 - 11 | 1582 | 77 | |
| XM_008259743.2 | XP_008257965.1 | 10 - 11 | 1576 | 77 | ||
| XM_017343405.1 | XP_017198894.1 | 9 - 10 | 1534 | 77 | ||
| XM_017343406.1 | XP_017198895.1 | 9 - 10 | 1486 | 77 | ||
| XM_017343407.1 | XP_017198896.1 | 8 - 9 | 1438 | 77 | ||
| XM_017343408.1 | XP_017198897.1 | 9 - 10 | 1398 | 77 | ||
| XM_017343409.1 | XP_017198898.1 | 8 - 9 | 1302 | 77 | ||
More Information
Set Membership: |
|

ZNF384
Genomic Map