Assay Details
| Gene Symbol: NPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714112.3 | XP_002714158.1 | 1 - 2 | 752 | 58 | |
| XM_008262100.2 | XP_008260322.1 | 1 - 2 | 752 | 58 | ||
More Information
Set Membership: |
|

NPR3
Genomic Map