Assay Details
Gene Symbol: LOC100343496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008269863.2 | XP_008268085.1 | 22 - 23 | 2494 | 86 | |
XM_008269864.2 | XP_008268086.1 | 22 - 23 | 2491 | 86 | ||
XM_008269865.2 | XP_008268087.1 | 22 - 23 | 2494 | 86 | ||
XM_008269866.2 | XP_008268088.1 | 22 - 23 | 2494 | 86 | ||
XM_008269867.2 | XP_008268089.1 | 22 - 23 | 2494 | 86 | ||
XM_017348677.1 | XP_017204166.1 | 21 - 22 | 2386 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
