Assay Details
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719045.3 | XP_002719091.1 | 4 - 5 | 649 | 65 | |
XM_008270924.2 | XP_008269146.1 | 2 - 3 | 174 | 65 | ||
XM_017348938.1 | XP_017204427.1 | 4 - 5 | 421 | 65 | ||
XM_017348939.1 | XP_017204428.1 | 4 - 5 | 421 | 65 | ||
XM_017348940.1 | XP_017204429.1 | 4 - 5 | 421 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008270924.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
