Assay Details
| Gene Symbol: LOC100339446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708640.3 | XP_002708686.1 | 14 - 15 | 1814 | 106 | |
| XM_017344075.1 | XP_017199564.1 | 14 - 15 | 1815 | 106 | ||
| XM_017344083.1 | XP_017199572.1 | 14 - 15 | 1625 | 106 | ||
More Information
Set Membership: |
|

LOC100339446
Genomic Map