Assay Details
Gene Symbol: LOC100349765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264416.2 | XP_008262638.1 | 1 - 2 | 102 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264416.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
