Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715207.3 | XP_002715253.1 | 3 - 4 | 461 | 62 | |
| XM_008264198.2 | XP_008262420.1 | 3 - 4 | 450 | 62 | ||
| XM_008264200.2 | XP_008262422.1 | 3 - 4 | 441 | 62 | ||
More Information
Set Membership: |
|

NIT1
Genomic Map