Assay Details
| Gene Symbol: MRTFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711759.3 | XP_002711805.1 | 10 - 11 | 2424 | 58 | |
| XM_002711760.3 | XP_002711806.2 | 12 - 13 | 2901 | 58 | ||
| XM_008257740.2 | XP_008255962.1 | 13 - 14 | 3066 | 58 | ||
| XM_008257741.2 | XP_008255963.1 | 14 - 15 | 3207 | 58 | ||
| XM_008257742.2 | XP_008255964.1 | 13 - 14 | 3042 | 58 | ||
| XM_008257744.2 | XP_008255966.1 | 10 - 11 | 2093 | 58 | ||
| XM_017342465.1 | XP_017197954.1 | 13 - 14 | 3143 | 58 | ||
More Information
Set Membership: |
|

MRTFB
Genomic Map