Assay Details
| Gene Symbol: HHATL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713051.3 | XP_002713097.2 | 7 - 8 | 1565 | 59 | |
| XM_017343669.1 | XP_017199158.1 | 8 - 9 | 1074 | 59 | ||
| XM_017343670.1 | XP_017199159.1 | 7 - 8 | 961 | 59 | ||
More Information
Set Membership: |
|

HHATL
Genomic Map