Assay Details
| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265483.2 | XP_008263705.1 | 1 | 2463 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CITED4| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265483.2 | XP_008263705.1 | 1 | 2463 | 94 | |
Set Membership: |
|