Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718145.3 | XP_002718191.1 | 5 - 6 | 1199 | 64 | |
| XM_017348265.1 | XP_017203754.1 | 5 - 6 | 1212 | 64 | ||
More Information
Set Membership: |
|

BRMS1L
Genomic Map