Assay Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711006.3 | XP_002711052.1 | 8 - 9 | 1341 | 106 | |
| XM_017341881.1 | XP_017197370.1 | 8 - 9 | 1366 | 106 | ||
More Information
Set Membership: |
|

KRT78
Genomic Map