Assay Details
Gene Symbol: LOC100343709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710450.3 | XP_002710496.1 | 3 - 4 | 403 | 72 | |
XM_008255595.2 | XP_008253817.1 | 3 - 4 | 403 | 72 | ||
XM_008255596.2 | XP_008253818.1 | 3 - 4 | 357 | 72 | ||
XM_008255598.2 | XP_008253820.1 | 3 - 4 | 388 | 72 | ||
XM_017341361.1 | XP_017196850.1 | 3 - 4 | 295 | 72 | ||
XM_017341362.1 | XP_017196851.1 | 3 - 4 | 192 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017341362.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
