Assay Details
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711601.2 | XP_002711647.1 | 9 - 10 | 1523 | 64 | |
XM_008257458.1 | XP_008255680.1 | 9 - 10 | 1523 | 64 | ||
XM_008257459.1 | XP_008255681.1 | 7 - 8 | 1349 | 64 | ||
XM_008257460.1 | XP_008255682.1 | 9 - 10 | 1523 | 64 | ||
XM_017342295.1 | XP_017197784.1 | 6 - 7 | 1201 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
