Assay Details
Gene Symbol: PRORP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717946.3 | XP_002717992.1 | 6 - 7 | 1904 | 72 | |
XM_008269496.2 | XP_008267718.1 | 7 - 8 | 2050 | 72 | ||
XM_008269497.2 | XP_008267719.1 | 7 - 8 | 2009 | 72 | ||
XM_017348261.1 | XP_017203750.1 | 5 - 6 | 554 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
