Assay Details
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713059.3 | XP_002713105.1 | 4 - 5 | 835 | 56 | |
XM_008260251.2 | XP_008258473.1 | 4 - 5 | 642 | 56 | ||
XM_008260252.2 | XP_008258474.1 | 4 - 5 | 1423 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
