Assay Details
| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719259.3 | XP_002719305.1 | 4 - 5 | 541 | 66 | |
| XM_008271200.2 | XP_008269422.1 | 4 - 5 | 541 | 66 | ||
| XM_008271201.2 | XP_008269423.1 | 4 - 5 | 542 | 66 | ||
More Information
Set Membership: |
|

GDPD1
Genomic Map