Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349037.1 | XP_017204526.1 | 3 - 4 | 486 | 57 | |
| GenBank mRNA | KF527431.1 | - | 2 - 3 | 187 | 57 | |
More Information
Set Membership: |
|

CCL5
Genomic Map