Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715521.3 | XP_002715567.2 | 5 - 6 | 1083 | 77 | |
XM_008264941.2 | XP_008263163.1 | 5 - 6 | 1087 | 77 | ||
XM_017346129.1 | XP_017201618.1 | 5 - 6 | 654 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
