Assay Details
| Gene Symbol: LOC100345689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714828.3 | XP_002714874.1 | 6 - 7 | 1303 | 89 | |
| XM_008263532.2 | XP_008261754.1 | 7 - 8 | 1557 | 89 | ||
| XM_008263535.2 | XP_008261757.1 | 7 - 8 | 1348 | 89 | ||
| XM_017345379.1 | XP_017200868.1 | 7 - 8 | 1560 | 89 | ||
More Information
Set Membership: |
|

LOC100345689
Genomic Map