Assay Details
| Gene Symbol: LOC100354826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350475.1 | XP_017205964.1 | 613 | 74 | ||
| XM_017350476.1 | XP_017205965.1 | 613 | 74 | |||
More Information
Set Membership: |
|

LOC100354826
Genomic Map