Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259665.2 | XP_008257887.1 | 3 - 4 | 480 | 76 | |
XM_008259666.2 | XP_008257888.1 | 2 - 3 | 743 | 76 | ||
XM_008259667.2 | XP_008257889.1 | 2 - 3 | 332 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
