Assay Details
Gene Symbol: AMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272710.2 | XP_008270932.1 | 2 - 3 | 329 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272710.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
