Assay Details
Gene Symbol: WAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272584.2 | XP_008270806.1 | 9 - 10 | 996 | 61 | |
XM_017349708.1 | XP_017205197.1 | 10 - 11 | 989 | 61 | ||
XM_017349709.1 | XP_017205198.1 | 10 - 11 | 1041 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
