Assay Details
| Gene Symbol: CEMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256353.2 | XP_008254575.1 | 6 - 7 | 1568 | 58 | |
| XM_008256356.2 | XP_008254578.1 | 6 - 7 | 1511 | 58 | ||
| XM_017341788.1 | XP_017197277.1 | 7 - 8 | 1621 | 58 | ||
| XM_017341798.1 | XP_017197287.1 | 7 - 8 | 1541 | 58 | ||
| XM_017341804.1 | XP_017197293.1 | 8 - 9 | 1712 | 58 | ||
| XM_017341810.1 | XP_017197299.1 | 6 - 7 | 1428 | 58 | ||
| XM_017341812.1 | XP_017197301.1 | 6 - 7 | 1573 | 58 | ||
| XM_017341820.1 | XP_017197309.1 | 6 - 7 | 1576 | 58 | ||
More Information
Set Membership: |
|

CEMIP2
Genomic Map