Assay Details
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249159.2 | XP_008247381.1 | 227 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249159.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
