Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345911.1 | XP_017201400.1 | 5 - 6 | 1940 | 66 | |
| XM_017345912.1 | XP_017201401.1 | 4 - 5 | 827 | 66 | ||
| XM_017345913.1 | XP_017201402.1 | 5 - 6 | 1028 | 66 | ||
More Information
Set Membership: |
|

CIART
Genomic Map