Assay Details
| Gene Symbol: MFGE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723373.3 | XP_002723419.3 | 389 | 61 | ||
| XM_008251720.2 | XP_008249942.2 | 389 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFGE8| Gene Symbol: MFGE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723373.3 | XP_002723419.3 | 389 | 61 | ||
| XM_008251720.2 | XP_008249942.2 | 389 | 61 | |||
Set Membership: |
|