Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711534.3 | XP_002711580.3 | 3 - 4 | 410 | 70 | |
XM_008257321.2 | XP_008255543.2 | 3 - 4 | 408 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
