Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265950.2 | XP_008264172.1 | 5 - 6 | 684 | 54 | |
| XM_008265951.2 | XP_008264173.1 | 5 - 6 | 708 | 54 | ||
| XM_008265957.2 | XP_008264179.1 | 4 - 5 | 587 | 54 | ||
More Information
Set Membership: |
|

NMNAT1
Genomic Map