Assay Details
| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255479.2 | XP_008253701.1 | 2 - 3 | 677 | 82 | |
| XM_008255480.2 | XP_008253702.1 | 3 - 4 | 754 | 82 | ||
| XM_008255481.2 | XP_008253703.1 | 3 - 4 | 796 | 82 | ||
| XM_008255482.2 | XP_008253704.1 | 2 - 3 | 1910 | 82 | ||
More Information
Set Membership: |
|

N4BP3
Genomic Map