Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714160.3 | XP_002714206.1 | 4 - 5 | 483 | 86 | |
| XM_017344878.1 | XP_017200367.1 | 5 - 6 | 460 | 86 | ||
More Information
Set Membership: |
|

LY86
Genomic Map